Få mere at vide
Baclofen Methyl Ester Hydrochloride, Mikromol™
Beskrivelse
Baclofen Methyl Ester Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Baclofen Methyl Ester Hydrochloride |
| CAS | 213407-11-5 |
| Analyt- eller komponentnavne | Baclofen Methyl Ester Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C11 H14 Cl N O2 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | β-(Aminomethyl)-4-chloro-benzenepropanoic acid methyl ester hydrochloride (1:1), Benzenepropanoic acid, β-(aminomethyl)-4-chloro-, methyl ester, hydrochloride (9CI) |
| InChI formel | InChI=1S/C11H14ClNO2.ClH/c1-15-11(14)6-9(7-13)8-2-4-10(12)5-3-8;/h2-5,9H,6-7,13H2,1H3;1H |
| SMIL | Cl.COC(=O)CC(CN)c1ccc(Cl)cc1 |
| IUPAC navn | methyl 4-amino-3-(4-chlorophenyl)butanoate;hydrochloride |
| Vis mere |