Få mere at vide
Baclofen-d4 (Major), TRC
Artikelnummer. 30341909
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30341909 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341909 Leverandør Toronto Research Chemicals Leverandørnr. TRCB10800310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Baclofen 2,4-butyl-d4
CAS 70155-52-1
Molekylær formel C10 2H4 H8 Cl N O2
Mængde 10 mg
Synonym β-(Aminomethyl-d2)-4-chlorobenzenepropanoic-α,α-d2 Acid
InChI formel InChI=1S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8H,5-6,12H2,(H,13,14)/i5D2,6D2
SMIL [2H]C([2H])(N)C(c1ccc(Cl)cc1)C([2H])([2H])C(=O)O
IUPAC navn 4-amino-3-(4-chlorophenyl)-2,2,4,4-tetradeuterio-butanoic acid
Molekylvægt (g/mol) 217.69
Formel vægt 217.0808