Få mere at vide
(±)-Baclofen-d4 (4-chlorophenyl-d4), CDN
Artikelnummer. 30316885
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316885 0.01 g
30316426 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316885 Leverandør CDN Leverandørnr. CDND75490.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Baclofen phenyl-d4
CAS 1189938-30-4
Molekylær formel C10 2H4 H8 Cl N O2
Mængde 0.01 g
Synonym β-(Aminomethyl)-4-chloro-benzenepropanoic Acid-d4, β-(Aminomethyl)-p-chlorohydrocinnamic Acid-d4, β-(4-Chlorophenyl)-GABA-d5, Ba-34647-d4, Baclon-d4, Clofen-d4, Lioresal-d4
InChI formel InChI=1 S/C10H12ClNO2/c11-9-3-1-7(2-4-9)8(6-12)5-10(13)14/h1-4,8 H,5-6,12H2,(H,13,14)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c(C(CN)CC(=O)O)c([2 H])c([2 H])c1Cl
IUPAC navn 4-amino-3-(4-chloro-2,3,5,6-tetradeuteriophenyl)butanoic acid
Molekylvægt (g/mol) 217.69
Formel vægt 217.0808 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity