missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ BACH1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17896642
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17896642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17896642 Leverandør Invitrogen™ Leverandørnr. PA5143055

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

BACH1.3 is a transcription factor that belongs to the cap'n'collar type of basic region leucine zipper factor family (CNC-bZip). The encoded protein contains broad complex, tramtrack, bric-a-brac/poxvirus and zinc finger (BTB/POZ) domains, which is atypical of CNC-bZip family members. These BTB/POZ domains facilitate protein-protein interactions and formation of homo and/or hetero oligomers. When this encoded protein forms a heterodimer with MafK, it functions as a repressor of Maf recognition element (MARE) and transcription is repressed. Multiple alternatively spliced transcript variants have been identified for this gene. Some exons of this gene overlap with some exons from the C21orf41 gene, which is transcribed in an opposite orientation to this gene, but which does not seem to encode a protein.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen BACH1
Applikationer Western Blot
Klassifikation Polyclonal
Koncentration 0.5 mg/mL
Konjugeret Unconjugated
Formulering TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene BACH1
Genadgangsnr. O14867
Gene Alias 6230421P05Rik; AI323795; BACH1; BACH-1; basic region leucine zipper transcriptional regulator BACH1; BTB and CNC homolog 1; BTB and CNC homology 1; BTB and CNC homology 1 transcription factor; BTB and CNC homology 1, basic leucine zipper transcription factor 1; BTB domain and CNC homolog 1; BTBD24; HA2303; Transcription regulator protein BACH1
Gensymboler BACH1
Værtsarter Goat
Immunogen Peptide with sequence HASVQPNERSENEC (aa 250-263).
Oprensningsmetode Ammonium sulfate precipitation
Mængde 100 μg
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 571
Målarter Human
Indhold og opbevaring -20°C, Avoid Freeze/Thaw Cycles
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.