missing translation for 'onlineSavingsMsg'
Få mere at vide

Baccatin VI, TRC

Artikelnummer. 30333208
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30333208 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30333208 Leverandør Toronto Research Chemicals Leverandørnr. TRCB10103050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Baccatin VI
CAS 57672-79-4
Molekylær formel C37 H46 O14
Mængde 50 mg
Synonym 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 4,5,6,9,12b-pentaacetate 12-benzoate, (2aR,4S,4aS,5R,6R,9S,11S,12S,12aR,12bS)-, 7,11-Methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-4,5,6,9,11,12,12b-heptol, 2a,3,4,4a,5,6,9,10,12,12a-decahydro-4a,8,13,13-tetramethyl-, 4,5,6,9,12b-pentaacetate 12-benzoate, [2aR-(2aα,4β,4aβ,5α,6β,9α,11α,12α,12aα,12bα)]-, Baccatin VI
InChI formel InChI=1S/C37H46O14/c1-18-25(46-19(2)38)16-37(44)32(50-33(43)24-13-11-10-12-14-24)30-35(9,26(47-20(3)39)15-27-36(30,17-45-27)51-23(6)42)31(49-22(5)41)29(48-21(4)40)28(18)34(37,7)8/h10-14,25-27,29-32,44H,15-17H2,1-9H3/t25-,26-,27+,29+,30-,31-,32-,35+,36-,37+/m0/s1
SMIL CC(=O)O[C@H]1C[C@@]2(O)[C@@H](OC(=O)c3ccccc3)[C@H]4[C@@](C)([C@H](C[C@H]5OC[C@@]45OC(=O)C)OC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)C(=C1C)C2(C)C
IUPAC navn [(1S,2S,3R,4S,7R,9S,10S,11R,12R,15S)-4,9,11,12,15-pentaacetyloxy-1-hydroxy-10,14,17,17-tetramethyl-6-oxatetracyclo[11.3.1.03,10.04,7]heptadec-13-en-2-yl] benzoate
Molekylvægt (g/mol) 714.75
Formel vægt 714.2888
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.