missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ B3GAT1 Polyclonal Antibody
GREENER_CHOICE

Artikelnummer. 17791344
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17791344 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 17791344 Leverandør Invitrogen™ Leverandørnr. PA5119947

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Rabbit Polyclonal Antibody

Positive Control: Rat brain tissue lysates, mouse brain tissue, SH-SY5Y. Subcellular Location: Secreted, golgi apparatus membrane, endoplasmic reticulum membrane.

The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen B3GAT1
Applikationer Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Koncentration 1 mg/mL
Konjugeret Unconjugated
Formulering TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene B3GAT1
Genadgangsnr. O35789, Q9CW73, Q9P2W7
Gene Alias 0710007K08Rik; AI846286; B3GAT1; Beta-1,3-glucuronyltransferase 1; beta-1,3-glucuronyltransferase 1 (glucuronosyltransferase P); CD57; Galactosylgalactosylxylosylprotein 3-beta-glucuronosyltransferase 1; GLCATP; glcAT-P; GLCUATP; GlcUAT-P; Glucuronosyltransferase P; glucuronosyltransferase-P; Hnk1; Hnk-1; LEU7; LEU7 antigen; NK1; NK-1; UDP-GlcUA:glycoprotein beta-1,3-glucuronyltransferase
Gensymboler B3GAT1
Værtsarter Rabbit
Immunogen Recombinant protein within human B3GAT1 aa 130-334 (Lumenal).
Oprensningsmetode Antigen affinity chromatography
Mængde 100 μL
Regulatorisk status RUO
Primær eller sekundær Primary
Gen-id (Entrez) 117108, 27087, 76898
Målarter Human, Mouse, Rat
Indhold og opbevaring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttype Antibody
Form Liquid
Isotype IgG
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.