Få mere at vide
BOLA B.safe Funnels 180
Beskrivelse
- Funnel made of PE / static dissipative PE-EX for liquid waste disposal
- The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck
- b.safe Funnels 180 are available in many versions
- The hinged lid respectively the ball valve reliably withhold vapours from the container
- The splash guard avoids spillover and splashing during funnel filling
- The static dissipative PE-EX version includes two attachable filling tubes
- Safe earthing of the funnel can be made via the grounding cable by qualified personnel
Tekniske data
Tekniske data
| Produkttype | Sikkerhedstragt |