Få mere at vide
BOLA B.safe Funnels 180
Artikelnummer. 17101206
Skift visning
Produkttype:
Sikkerhedstragt
Artikelnummer. Produkttype
17101206 Sikkerhedstragt
1 options
Artikelnummer. 17101206 Leverandør BOLA Leverandørnr. R84010
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Made of static dissipative PE-EX conductive, with sieve, cover and grounding cable 1.5 m
  • Funnel made of PE / static dissipative PE-EX for liquid waste disposal
  • The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with the filling neck
  • b.safe Funnels 180 are available in many versions
  • The hinged lid respectively the ball valve reliably withhold vapours from the container
  • The splash guard avoids spillover and splashing during funnel filling
  • The static dissipative PE-EX version includes two attachable filling tubes
  • Safe earthing of the funnel can be made via the grounding cable by qualified personnel
Tekniske data
Produkttype Sikkerhedstragt