Få mere at vide
Tocris Bioscience™ B I09
GREENER_CHOICE
IRE1 endonuclease inhibitor; cell permeable
Brand:  Tocris Bioscience™ 6009/50
Encompass
Produktkode 15767186
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
B I09 is an IRE-1 RNase inhibitor (IC50 = 1.23 μM). Inhibits IRE-1/XBP1 pathway. Cell permeable. Inhibits growth of human chronic lymphocytic leukemia (CLL) cells in vitro and promotes CLL regression in a mouse model.
Tekniske data
Tekniske data
Beige
1607803-67-7
303.31g/mol
Store at -20°C
50 mg
Inhibitor
C16H17NO5
Soluble to 50mM in DMSO
Inhibits Rnase
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only