missing translation for 'onlineSavingsMsg'
Få mere at vide

b-Chloro-D-alanine Hydrochloride, TRC

Artikelnummer. 30350229
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30350229 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30350229 Leverandør Toronto Research Chemicals Leverandørnr. TRCC364200250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale beta-Chloro-D-alanine hydrochloride
CAS 51887-88-8
Molekylær formel C3 H6 Cl N O2 . Cl H
Mængde 250 mg
Synonym D-Alanine, 3-chloro-, hydrochloride (1:1), D-Alanine, 3-chloro-, hydrochloride (9CI), 3-Chloro-D-alanine hydrochloride, D-β-Chloroalanine hydrochloride, β-Chloro-D-alanine hydrochloride, (2S)-2-Amino-3-chloro-propanoic acid hydrochloride
InChI formel InChI=1S/C3H6ClNO2.ClH/c4-1-2(5)3(6)7;/h2H,1,5H2,(H,6,7);1H/t2-;/m1./s1
SMIL Cl.N[C@H](CCl)C(=O)O
IUPAC navn (2S)-2-amino-3-chloropropanoic acid;hydrochloride
Molekylvægt (g/mol) 160
Formel vægt 158.9854
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.