Få mere at vide
B-[2-Fluoro-3-(methoxy-d3)phenyl]boronic Acid, TRC
Artikelnummer. 30406782
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30406782 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30406782 Leverandør Toronto Research Chemicals Leverandørnr. TRCF40299625MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale B-[2-Fluoro-3-(methoxy-d3)phenyl]boronic Acid
CAS 2233595-37-2
Molekylær formel C7H5D3BFO3
Mængde 25 mg
InChI formel InChI=1S/C7H8BFO3/c1-12-6-4-2-3-5(7(6)9)8(10)11/h2-4,10-11H,1H3/i1D3
SMIL [2H]C([2H])([2H])Oc1cccc(B(O)O)c1F
IUPAC navn [2-fluoro-3-(trideuteriomethoxy)phenyl]boronic acid
Molekylvægt (g/mol) 172.96
Formel vægt 173.0739