missing translation for 'onlineSavingsMsg'
Learn More

Azure I, Thermo Scientific Chemicals

Catalog Number p-7023694
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
531-55-5
C15H16ClN3S
305.824
MFCD00011935
DNDJEIWCTMMZBX-UHFFFAOYSA-N
azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech
68275
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]
This item is not returnable. View return policy
531-55-5
C15H16ClN3S
305.824
MFCD00011935
DNDJEIWCTMMZBX-UHFFFAOYSA-N
azure b, azure i, methylene azure b, 3-dimethylamino-7-methylamino phenothiazin-5-ium chloride, trimethylthionine chloride, azur i, methyleneazure, methylene azure van, unii-7c8bel8wqv, modr kresylova bb czech
68275
dimethyl-[7-(methylamino)phenothiazin-3-ylidene]azanium;chloride
CNC1=CC2=C(C=C1)N=C3C=CC(=[N+](C)C)C=C3S2.[Cl-]

Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼205°C (decomposition)
Odor Odorless
Quantity 5 g
Beilstein 3922630
Merck Index 14,6058
Solubility Information Soluble in 1mg/ml water. Sparingly soluble in alcohol.
Formula Weight 305.83
Chemical Name or Material Azure I

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'