missing translation for 'onlineSavingsMsg'
Få mere at vide

Azorubin (E122) 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Artikelnummer. 30316650
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30316650 1 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30316650 Leverandør Dr. Ehrenstorfer Leverandørnr. DREA10411500AL100

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Spécification

Kemisk navn eller materiale Azorubin (E122)
CAS 3567-69-9
Analyt- eller komponentnavne Azorubin (E122)
Forsendelsestilstand Room Temperature
Molekylær formel C20 H12 N2 O7 S2 . 2 Na
Koncentration 100 μg/mL
Certificeringer/compliance ISO 17034
Synonym 4-Hydroxy-3-[2-(4-sulfo-1-naphthalenyl)diazenyl]-1-naphthalenesulfonic Acid Sodium Salt (1:2);1-Naphthalenesulfonic acid, 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-, disodium salt (9CI), Azo Rubine (6CI), C.I. Acid Red 14, disodium salt (8CI), 11959 Red, AR 14, Acid Brilliant Rubine 2G, Acid Carmosine S, Acid Chrome Blue 2R, Acid Chrome Blue BA, Acid Chrome Blue BA-CF, Acid Chrome Blue FBS, Acid Fast Red FB, Acid Naphthol Red B, Acid Red 14, Acid Red 2S, Acid Red B, Acid Red B (Chinese), Acid Rubine, Acid Violet Red, Acidic Red B, Airedale Carmoisine, Amacid Carmoisine B, Amacid Chrome Blue R, Amaranth 36028, Azo Rubin Extra, Azo Rubine LZ, Azo Rubine S, Azo Rubine S Specially Pure, Azo Rubine XX, Azo Rubine for Food, Azorubin, Basovit Red 440E, Brasilan Azo Rubine 2NS, Brilliant Acid Rubine M, Brilliant Carmoisine, Brilliant Crimson 2R-FQ, Brilliant Crimson Red, Bucacid Azo Rubine, C.I. 14720, C.I. Acid Red 14, C.I. Food Red 3, C.I. Mordant Blue 79, Calcocid Rubine XX, Carmoisine, Carmoisine 206, Carmoisine 36011, Carmoisine 36025, Carmoisine 90141, Carmoisine A, Carmoisine B, Carmoisine BA-CF, Carmoisine GRN, Carmoisine LAS, Carmoisine S, Carmoisine W, Carmoisine WNN 19153, Carmoisine azorubine, Certicol Carmoisine S, Chrome Fast Blue 2R, Chromotrope FB, Cilefa Rubine R, Cogilor Red 319.11, Cogilor Red 319.11-90022, Colorosacid Red 2B, Conacid Red AD, Crimson 2EMBL, Crimson EMBL, Diadem Chrome Blue G, Diadem Chrome Blue R, Dinacid Crystal Red A, Disodium 2-(4-sulfo-1-naphthylazo)-1-naphthol-4-sulfonate, Dyacid Red W, Dycosacid Red B, Dynacid Red C, E 122, Edicol Supra Carmoisine W, Edicol Supra Carmoisine WS, Eniacid Brilliant Rubine 3B, Erio Rubine B, Eurocert Azorubine, Eurocert Carmoisine 311804, Ext D and C Red No. 10, Fenazo Red C, Food red 3, Fruit Red A Extra Yellowish Geigy, HD Carmoisine, HD Carmoisine Supra, Hexacol Carmoisine, Hidacid Azo Rubine, Hispacid Rubine F, Java Rubine N, Karmesin, Kemacid Carmosine WS, Kenachrome Blue 2R, Kiton Crimson 2R, Kiton Rubine R, Lighthouse Chrome Blue 2R, Lissamine Red W, Nacarat, Nacarat A Export, Nacarat Extra Pure A, Necol Carmoisine, Neelicol Carmoisine, Neklacid Azorubine W, Omega Chrome Blue FB, Pontacyl Rubine R, PuriColor Red ARE 14, Rubine B, Sicovit Azorubine 85E122, Solar Rubine, Solochrome Blue FB, Tertracid Red CA, Tertrochrome Blue FB, Triacid Rubine WS, Water Red 176554, Water Red 176573
InChI formel InChI=1S/C20H14N2O7S2.2Na/c23-20-15-8-4-3-7-14(15)19(31(27,28)29)11-17(20)22-21-16-9-10-18(30(24,25)26)13-6-2-1-5-12(13)16;;/h1-11,23H,(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2
SMIL [Na+].[Na+].Oc1c(cc(c2ccccc12)S(=O)(=O)[O-])N=Nc3ccc(c4ccccc34)S(=O)(=O)[O-]
IUPAC navn disodium;4-hydroxy-3-[(E)-(4-sulfonatonaphthalen-1-yl)diazenyl]naphthalene-1-sulfonate
Opløsningsmiddel eller Matrix Acetonitrile
Molekylvægt (g/mol) 502.43
Mængde 1 ml
Formel vægt 501.9881 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.