Få mere at vide
Azlon™ Tilspidset glasfuge
Standard konusled nr.:
10/19
14/23
19/26
24/29
29/32
34/35
Pakningsstørrelse:
Hver
Beskrivelse
- For grease free and high vacuum seals using Quickfit™ ground glass joints
- Strong construction with knurled shoulder for ease of handling and repeated use
- Self lubricating and non-contaminating
- Working temperature range -100°C to +260°C
- Working vacuum ≤10-3atmospheres
- Highly resistant to chemical attack
Tekniske data
Tekniske data
| Materiale | PTFE |
| Produkttype | Tapered sleeves |
| Mængde | 1/Pk. |
| Standard konusled nr. | 34/35 |