missing translation for 'onlineSavingsMsg'
Learn More

Azlon Polypropylene T Pattern Connector

Brand:  Azlon CWH210

 View more versions of this product

Product Code. 10136354

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Tapered arms are rigid with steps to give firm grip
  • Autoclavable at 121°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
Polypropylene
T Pattern
Connector
9 mm
10/Pk.
White
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon Polypropylene T Pattern Connector > I.D.: 9mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.