missing translation for 'onlineSavingsMsg'
Få mere at vide

Azithromycin dihydrate, Thermo Scientific™

Azithromycin dihydrate, CAS # 117772-70-0, is an azalide derived from erythromycin that shows antibacterial activity.

Brand:  Thermo Scientific Alfa Aesar J66740.06

2006.00 DKK valid until 2026-07-03
BEDSTE tilbudspris! Brug kampagnekoden "27965" for at få din tilbudspris.



Mængde 5g

Produktkode 15498209

  • ONLINE EXCLUSIVE PRICE
    2865.71 DKK
    2578.65 DKK / 5g
    Spare 287.06 DKK
    10% Slukket
Se alle kampagner

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

General Description

• Azithromycin dihydrate is the dihydrate form of azithromycin, an azalide derived from erythromycin, that shows antibacterial activity

• Azithromycin dihydrate reversibly binds to the 23S rRNA of the 50S ribosomal subunit of the bacterial ribosome of susceptible microorganisms, inhibiting the translocation step of protein synthesis by preventing the assembly of the 50S ribosomal subunit. This inhibits bacterial protein synthesis and cell growth, causing cell death

Application

• Azithromycin dihydrate shows anti-immunomodulatory and anti-inflammatory activities

• It shows antimalarial activity against Plasmodium falciparum and P. vivax, which are multidrug-resistant species

• In the laboratory, azithromycin dihydrate has been used in blocking buffer for indirect immunofluorescence

• It has been used in U87 cells for cell viability assay and flow cytometry analysis to study the Zika virus infection in glial cells

• Azithromycin dihydrate has been used in experiments to compare its antibacterial activity with PYRRO-C3D against Streptococcus pneumoniae biofilms

TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Azithromycin dihydrate
White
>110°C (230°F)
C38H76N2O14
717°C
Soluble in DMSO at 20mg/mL; Also soluble in chloroform or ethylene chloride
CCC1C(C(C(N(CC(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)O)C)C)C)O)(C)O.O.O
785.026
785.01
117772-70-0
5g
Air sensitive
MFCD01862248
N-Methyl-11-aza-10-deoxo-10-dihydroerythromycin A; CP-62993
SRMPHJKQVUDLQE-GTCGPNQGSA-N
(2R,3S,4S,5R,8R,10R,11R,12S,13R,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-o
131845180
98%
Produktforslag

Produktforslag

Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

Produktidentifikator
  • Azithromycin dihydrate

Signalord
  • Fare

Farekategori
  • Respiratorisk sensibiliserende stof Kategori 1
  • Hudsensibilisering Kategori 1

Faresætning
  • H317-Kan forårsage allergisk hudreaktion.
  • H334-Kan forårsage allergi- eller astmasymptomer eller åndedrætsbesvær ved indånding.

Sikkerhedssætning
  • P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
  • P284-[I tilfælde af utilstrækkelig ventilation], anvend åndedrætsværn.
  • P302+P352-VED KONTAKT MED HUDEN: Vask med rigeligt vand/.
  • P304+P340-VED INDÅNDING: Flyt personen til et sted med frisk luft og sørg for, at vejrtrækningen lettes.
  • P333+P313-Ved hudirritation eller udslet: Søg lægehjælp.
  • P342+P311-Ved luftvejssymptomer: Ring til en GIFTINFORMATION/læge/ .
  • P362+P364-Alt tilsmudset tøj tages af og vaskes inden genanvendelse.

Supplerende oplysninger
  • MIXTURE LIST-Contain: Azithromycin hydrate

SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.