missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Azinphos-ethyl D10 100 μg/mL in Acetone, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Azinphos-ethyl D10 (ethyl D10) |
| CAS | 2732916-08-2 |
| Opløsningsmiddel eller Matrix | Acetone |
| Molekylær formel | C12 2H10 H6 N3 O3 P S2 |
| Mængde | 1 mL |
| Synonym | Azinphos-ethyl D10 (ethyl D10) |
| InChI formel | InChI=1S/C12H16N3O3PS2/c1-3-17-19(20,18-4-2)21-9-15-12(16)10-7-5-6-8-11(10)13-14-15/h5-8H,3-4,9H2,1-2H3/i1D3,2D3,3D2,4D2 |
| SMIL | [2H]C([2H])([2H])C([2H])([2H])OP(=S)(OC([2H])([2H])C([2H])([2H])[2H])SCN1N=Nc2ccccc2C1=O |
| IUPAC navn | 3-[bis(1,1,2,2,2-pentadeuterioethoxy)phosphinothioylsulfanylmethyl]-1,2,3-benzotriazin-4-one |
| Molekylvægt (g/mol) | 355.44 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?