Få mere at vide
Azilsartan Methyl Ester, TRC
Artikelnummer. 30135314
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135314 500 mg
30006585 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135314 Leverandør Toronto Research Chemicals Leverandørnr. TRCA926905500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 147403-52-9
Molekylær formel C26 H22 N4 O5
Molekylvægt (g/mol) 470.482
Synonym Methyl 2-Ethoxy-1-[[2'-(2,5-dihydro-5-oxo-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]methyl]benzimidazole-7-carboxylate, Methyl 2-Ethoxy-1-[[2'-(5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)biphenyl-4-yl]methyl]-1H-benzimidazole-7-carboxylate;
IUPAC navn methyl 2-ethoxy-3-[[4-[2-(5-oxo-4H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate
SMIL CCOc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C5=NOC(=O)N5
Tekniske data
Kemisk navn eller materiale Azilsartan Methyl Ester
Analyt- eller komponentnavne Azilsartan Methyl Ester
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C26H22N4O5/c1-3-34-25-27-21-10-6-9-20(24(31)33-2)22(21)30(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23-28-26(32)35-29-23/h4-14H,3,15H2,1-2H3,(H,28,29,32)
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 500 mg
Formel vægt 470.159
Procent renhed >95%