missing translation for 'onlineSavingsMsg'
Få mere at vide

Azilsartan Dimer, TRC

Artikelnummer. 30338492
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338492 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30338492 Leverandør Toronto Research Chemicals Leverandørnr. TRCA926920500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Azilsartan Dimer
CAS 1604812-35-2
Molekylær formel C35H28N4O11
Mængde 500 mg
Synonym (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-1-((2'-(4-((5-methyl-2-oxo-1,3-dioxol-4-yl)methyl)-5-oxo-4,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-benzo[d]imidazole-7-carboxylate, Azilsartan Impurity 9
InChI formel InChI=1S/C35H28N4O11/c1-4-44-32-36-26-11-7-10-25(31(40)45-18-28-20(3)47-35(43)49-28)29(26)38(32)16-21-12-14-22(15-13-21)23-8-5-6-9-24(23)30-37-50-33(41)39(30)17-27-19(2)46-34(42)48-27/h5-15H,4,16-18H2,1-3H3
SMIL CCOc1nc2cccc(C(=O)OCC3=C(C)OC(=O)O3)c2n1Cc4ccc(cc4)c5ccccc5C6=NOC(=O)N6CC7=C(C)OC(=O)O7
IUPAC navn (5-methyl-2-oxo-1,3-dioxol-4-yl)methyl 2-ethoxy-3-[[4-[2-[4-[(5-methyl-2-oxo-1,3-dioxol-4-yl)methyl]-5-oxo-1,2,4-oxadiazol-3-yl]phenyl]phenyl]methyl]benzimidazole-4-carboxylate
Molekylvægt (g/mol) 680.62
Formel vægt 680.1755
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.