Få mere at vide
Azilsartan-d5 Methyl Ester, TRC
Artikelnummer. 30336486
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30336486 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30336486 Leverandør Toronto Research Chemicals Leverandørnr. TRCA9269071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Azilsartan-d5 Methyl Ester
CAS 1794766-42-9
Molekylær formel C26H17D5N4O5
Mængde 1 mg
Synonym 1-[[2'-(2,5-Dihydro-5-oxo-1,2,4-oxadiazol-3-yl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic-d5 Acid Methyl Ester
InChI formel InChI=1S/C26H22N4O5/c1-3-34-25-27-21-10-6-9-20(24(31)33-2)22(21)30(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23-28-26(32)35-29-23/h4-14H,3,15H2,1-2H3,(H,28,29,32)/i1D3,3D2
SMIL [2H]C([2H])([2H])C([2H])([2H])Oc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C5=NC(=O)ON5
IUPAC navn methyl 3-[[4-[2-(5-oxo-2H-1,2,4-oxadiazol-3-yl)phenyl]phenyl]methyl]-2-(1,1,2,2,2-pentadeuterioethoxy)benzimidazole-4-carboxylate
Molekylvægt (g/mol) 475.51
Formel vægt 475.1904