Learn More
Azepan-2-one, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 105-60-2 |
| Molecular Formula | C6H11NO |
| MDL Number | MFCD00006936 |
| Quantity | 1 kg |
| SMILES | O=C1CCCCCN1 |
| Molecular Weight (g/mol) | 113.16 |
| Percent Purity | 99% |
| Chemical Name or Material | Azepan-2-one |