Få mere at vide
AZD9056 hydrochloride, MedChemExpress
Artikelnummer. 16490228
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16490228 5 mg
16447768 10 mg
16413737 50 mg
16457616 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16490228 Leverandør Medchem Express Leverandørnr. HY19427A
MedChemExpress AZD9056 hydrochloride is a selective orally active inhibitor of P2X7 which plays a significant role in inflammation and pain-causing diseases.
AZD9056 hydrochloride is a selective orally active inhibitor of P2X7 which plays a significant role in inflammation and pain-causing diseases.
Tekniske data
Kemisk navn eller materiale AZD9056 hydrochloride
Fysisk form Solid
CAS 345303-91-5
Farve White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C24H36Cl2N2O2
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : 50 mg/mL (109.78 mM; Need ultrasonic) ∣H2O : 1.67 mg/mL (3.67 mM; ultrasonic and warming and heat to 60°C)
SMIL O=C(C1=CC(CCCNCCCO)=CC=C1Cl)NCC2(C[C@H](C3)C4)C[C@H]4C[C@H]3C2.Cl
Molekylvægt (g/mol) 455.46
Formel vægt 455.46
Procent renhed 98.14%