Få mere at vide
AZD3839, TRC
Artikelnummer. 30338068
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30338068 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338068 Leverandør Toronto Research Chemicals Leverandørnr. TRCA80814525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale AZD3839
CAS 1227163-56-5
Molekylær formel C24H15F4N5
Mængde 25 mg
Synonym 4-Fluoro-1-[3-(5-pyrimidinyl)phenyl]-1-[2-(trifluoromethyl)-4-pyridinyl]-1H-isoindol-3-amine
InChI formel InChI=1S/C24H15F4N5/c25-19-6-2-5-18-21(19)22(29)33-23(18,17-7-8-32-20(10-17)24(26,27)28)16-4-1-3-14(9-16)15-11-30-13-31-12-15/h1-13H,(H2,29,33)
SMIL Cc1ccc(cc1)c2c(CN(C)C)n3c(ccc(C)c3)n2
IUPAC navn 7-fluoro-3-(3-pyrimidin-5-ylphenyl)-3-[2-(trifluoromethyl)pyridin-4-yl]isoindol-1-amine
Molekylvægt (g/mol) 449.4