Få mere at vide
Tocris Bioscience™ AZD 1480
GREENER_CHOICE
Potent and selective JAK2 inhibitor; antiangiogenic
Brand:  Tocris Bioscience™ 5617/10
Encompass
Produktkode 15716986
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AZD 1480 is a potent and selective JAK2 inhibitor (IC50 < 3 nM). Exhibits >50-fold selectivity for JAK2 over JAK3. Exhibits antitumor effects and inhibits formation of lung metastases in a mouse renal cancer model. Also inhibits STAT3 signaling, tumor angiogenesis and myeloid cell migration in vivo.
Tekniske data
Tekniske data
5-Chloro-N2-[(1S)-1-(5-fluoro-2-pyrimidinyl)ethyl]-N4-(5-methyl-1H-pyrazol-3-yl)-2,4-pyrimidine-2,4-diamine
935666-88-9
10 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner