Få mere at vide
AZD-1480, MedChemExpress
Artikelnummer. 16494726
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
Pakningsstørrelse:
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16494726 100 mg -
16470037 50 mg -
16456377 5 mg 5mg
16433648 10 mg -
16430946 200 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16494726 Leverandør Medchem Express Leverandørnr. HY10193
MedChemExpress AZD-1480 is an ATP-competitive inhibitor of JAK1 and JAK2 with IC50s of 1.3 nM and <0.4 nM, respectively.
AZD-1480 is an ATP-competitive inhibitor of JAK1 and JAK2 with IC50s of 1.3 nM and <0.4 nM, respectively.
Tekniske data
Kemisk navn eller materiale AZD-1480
Fysisk form Solid
CAS 935666-88-9
Farve White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C14H14ClFN8
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : 50 mg/mL (143.36 mM; Need ultrasonic)
SMIL ClC1=C(N=C(N=C1)N[C@@H](C)C2=NC=C(C=N2)F)NC3=NNC(C)=C3
Molekylvægt (g/mol) 348.77
Formel vægt 348.77
Procent renhed 98.02%