Få mere at vide
Tocris Bioscience™ AZD 1283
GREENER_CHOICE
High affinity P2Y12 antagonist
Brand:  Tocris Bioscience™ 6085/50
Encompass
Produktkode 15797206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
AZD 1283 is a potent and high affinity P2Y12 antagonist (IC50 = 11 nM in radioligand competition binding assay, and 25 nM in a GTPγS binding assay). Causes a dose-dependent increase in blood flow and inhibits ADP-induced platelet aggregation in vivo. Antithrombotic.
Tekniske data
Tekniske data
Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate
Ethyl 5-cyano-2-methyl-6-[4-[[[(phenylmethyl)sulfonyl]amino]carbonyl]-1-piperidinyl]-3-pyridinecarboxylate
Soluble to 100mM in DMSO
White
919351-41-0
50 mg
Solid
Bioactive Small Molecule
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only.