Få mere at vide
Azasetron-d3 Hydrochloride, TRC
Artikelnummer. 30338413
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30338413 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30338413 Leverandør Toronto Research Chemicals Leverandørnr. TRCA80280210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Azasetron-d3, Hydrochloride
CAS 1216505-58-6
Molekylær formel C17 2H3 H17 Cl N3 O3 . Cl H
Mængde 10 mg
InChI formel InChI=1S/C17H20ClN3O3.ClH/c1-20-14-7-11(18)6-12(16(14)24-9-15(20)22)17(23)19-13-8-21-4-2-10(13)3-5-21;/h6-7,10,13H,2-5,8-9H2,1H3,(H,19,23);1H/i1D3;
SMIL Cl.[2H]C([2H])([2H])N1C(=O)COc2c1cc(Cl)cc2C(=O)NC3CN4CCC3CC4
IUPAC navn N-(1-azabicyclo[2.2.2]octan-3-yl)-6-chloro-3-oxo-4-(trideuteriomethyl)-1,4-benzoxazine-8-carboxamide;hydrochloride
Molekylvægt (g/mol) 389.29
Formel vægt 388.1148