Få mere at vide
Azasetron-13C,D3 Hydrochloride, TRC
Artikelnummer. 30331502
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30331502 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331502 Leverandør Toronto Research Chemicals Leverandørnr. TRCA8028035MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Azasetron-13C,D3 Hydrochloride
CAS 1329611-23-5
Molekylær formel C1613CH18D3Cl2N3O3
Mængde 5 mg
Synonym N-1-Azabicyclo[2.2.2]-oct-3-yl-6-chloro-3,4-dihydro-4-(methyl-13C,d3)-3-oxo-2H-1,4-benzoxazine-8-carboxamide Hydrochloride
InChI formel InChI=1S/C17H20ClN3O3.ClH/c1-20-14-7-11(18)6-12(16(14)24-9-15(20)22)17(23)19-13-8-21-4-2-10(13)3-5-21;/h6-7,10,13H,2-5,8-9H2,1H3,(H,19,23);1H/i1+1D3;
SMIL Cl.[2H][13C]([2H])([2H])N1C(=O)COc2c1cc(Cl)cc2C(=O)NC3CN4CCC3CC4
IUPAC navn N-(1-azabicyclo[2.2.2]octan-3-yl)-6-chloro-3-oxo-4-(trideuterio(1^{13}C)methyl)-1,4-benzoxazine-8-carboxamide;hydrochloride
Molekylvægt (g/mol) 390.28
Formel vægt 389.1182