missing translation for 'onlineSavingsMsg'
Learn More

Azaperone, Thermo Scientific Chemicals

Catalog Number p-7077856
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
1649-18-9
C19H22FN3O
327.40
MFCD00866692
XTKDAFGWCDAMPY-UHFFFAOYSA-N
azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum
15443
1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1
This item is not returnable. View return policy
1649-18-9
C19H22FN3O
327.40
MFCD00866692
XTKDAFGWCDAMPY-UHFFFAOYSA-N
azaperone, stresnil, fluoperidol, suicalm, azaperon, azeperone, eucalmyl, sedaperone vet, azaperona, azaperonum
15443
1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-one
FC1=CC=C(C=C1)C(=O)CCCN1CCN(CC1)C1=CC=CC=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Loss on Drying 0.5% max. (60°C, 4 h) (vacuum)
Infrared Spectrum Authentic
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol, chloroform
Formula Weight 327.4
Chemical Name or Material Azaperone
compliance-icons
Product Identifier
  • Azaperone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'