Få mere at vide
Azaperol, TRC
Artikelnummer. 17932635
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17932635 25 mg
17903825 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17932635 Leverandør Toronto Research Chemicals Leverandørnr. TRCA80219025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2804-05-9
Molekylær formel C19 H24 F N3 O
Molekylvægt (g/mol) 329.41
Synonym 1-Piperazinebutanol, α-(p-fluorophenyl)-4-(2-pyridyl)- (6CI,8CI), α-(4-Fluorophenyl)-4-(2-pyridinyl)-1-piperazinebutanol, Azaperol, Dihydroazaperone
IUPAC navn 1-(4-fluorophenyl)-4-(4-pyridin-2-ylpiperazin-1-yl)butan-1-ol
SMIL OC(CCCN1CCN(CC1)c2ccccn2)c3ccc(F)cc3
Tekniske data
Kemisk navn eller materiale Azaperol
Analyt- eller komponentnavne Azaperol
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C19H24FN3O/c20-17-8-6-16(7-9-17)18(24)4-3-11-22-12-14-23(15-13-22)19-5-1-2-10-21-19/h1-2,5-10,18,24H,3-4,11-15H2
Løsningstype Neat
Industry Type Forensics
Mængde 25 mg
Formel vægt 329.1903
Procent renhed >95%