Få mere at vide
Azamulin, TRC
Artikelnummer. 30012883
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30012883 25 mg
30003644 10 mg
30009860 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30012883 Leverandør Toronto Research Chemicals Leverandørnr. TRCA80153025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76530-44-4
Molekylær formel C24 H38 N4 O4 S
Tekniske data
Kemisk navn eller materiale Azamulin
Mængde 25 mg
Forsendelsestilstand Room Temperature
Formel vægt 478.2614
Produkttype Bioactive Small Molecule
Renhed >95% (HPLC)
Molekylvægt (g/mol) 478.65
InChI nøgle FMHQJXGMLMSMLC-WBUYAQKGSA-N
SMIL [H][C@@]12C(=O)CC[C@]11CC[C@@H](C)[C@@]2(C)[C@@H](C[C@@](C)(CC)[C@@H](O)[C@@H]1C)OC(=O)CSC1=NNC(N)=N1
IUPAC navn (1S,2R,3S,4R,6R,7R,8R,14R)-4-ethyl-3-hydroxy-2,4,7,14-tetramethyl-9-oxotricyclo[5.4.3.0¹,⁸]tetradecan-6-yl 2-[(5-amino-1H-1,2,4-triazol-3-yl)sulfanyl]acetate