Få mere at vide
Tocris Bioscience™ AZ 11645373
GREENER_CHOICE
Artikelnummer. 18064962
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18064962 10 mg
18034962 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18064962 Leverandør Tocris Bioscience™ Leverandørnr. 3317/10
Potent and selective human P2X7 antagonist
Potent and selective human P2X7 antagonist (KB values are 5 - 7 and > 10,000 nM at hP2X7 and rP2X7 respectively) that is completely without effect at all other P2X subtypes. Inhibits BzATP-mediated calcium influx and inhibits ATP-mediated IL-1β release in vitro (KB values are 15 and 92 nM respectively).
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 227088-94-0
Molekylær formel C24H21N3O5S
Tekniske data
Kemisk navn eller materiale AZ 11645373
Mængde 10 mg
Formel vægt 463.51
Renhed >98%
Molekylvægt (g/mol) 463.51