Learn More
Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices
Ensure consistent performance and quality. Axygen™ Automation Tips for Tecan™, Zymark™, Biotek™ and Dynamic Devices are constructed of clear polypropylene in 96-well racks. Available in nonsterile/sterile, non-filtered/filtered, wide-bore and bottom-labeled options with volumes ranging from 50 to 200μL.
Brand:  Axygen™ EV-50-R-S
Additional Details : Weight : 9.50000kg
 View more versions of this product
Product Code. 12792836
  • 4023.00 DKK / Pack of 4800
Estimated Shipment
21-04-2021
Add to basket
Description
Description
  • Comprehensive line of pipette tips for automated liquid handling systems
  • Manufactured to stringent specifications under strict process controls
  • Assembled via automation to ensure consistent performance and quality
  • Tips are certified RNase-/DNase-free and nonpyrogenic
  • Available in a range of volumes: 50, 100, 180 and 200μL
  • Compatible with the following:
    • Tecan™ Freedom Evo™, TeMo and Aquarius with MCA96 head
    • Zymark™/Caliper RapidPlate™, SciClone™ and Zephyr™ with HV96 head
    • Biotek™ Precision™ and Precision XS 8 or 12 head
    • Dynamic Devices Oasis LM 8/96-Channel-Standard Vol and 96-Channel-Low Vol head
Specifications
Specifications
50 μL
Sterile
Light Green
PP (Polypropylene)
DNase, Rnase Free
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius, Zymark™/Caliper™ RapidPlate™, SciClone™, Zephyr™, BioTek™ Precision™, BioTek™ Precision™ XS, Dynamic Devices Oasis LM
Rack
SBS Rack
Non-Filtered
Clear
Round
ANSI
Non-pyrogenic, used for stringent specifications under strict process controls
Tecan™ Freedom EVO™, Tecan™ TeMo, Tecan™ Aquarius, Zymark™/Caliper™ RapidPlate™, SciClone™, Zephyr™, BioTek™ Precision™, BioTek™ Precision™ XS, Dynamic Devices Oasis LM
Automation
50 Racks/Cs.