Få mere at vide
Axygen™ 1,5 mL koniske rør med skruelåg
Med kemikaliebestandig O-ring
Brand: Axygen™ SCT-150-V
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order
- Caps available in range of colors for easy sample identification
Tekniske data
RNase, DNase, pyrogen-free | |
Polypropylene | |
Polypropylene | |
O-ring | |
No | |
Conical |
Violet | |
1.5 mL | |
Non-looped | |
Non-sterile | |
Screw Cap | |
20,000 x g |
Korrektion af produktindhold
Produkttitel
Axygen™ 1,5 mL koniske rør med skruelåg > Color: Violet; Nonsterile
Ser du en mulighed for forbedring?Del en indholdskorrektion