Få mere at vide
Axygen™ 0,5 mL Selvstående rør med skruelåg
Artikelnummer. 11330355 Shop alle Axygen Scientific produkter
Sterilitet:
Ikke steril
Ikke-steril
Steril
Lukningsfarve:
Blå
Brun
Diverse
Grøn
Grå
Gul
Hvid
Klar
Orange
Rød
Violet
Pakningsstørrelse:
Pakke med 4000
Pakke med 500
Product Code. 11330355
Brand: Axygen™ SCT050SSA
Har O-ring fremstillet til at være modstandsdygtig over for mange almindeligt anvendte laboratoriekemikalier
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
  • Available as presterilized and nonsterile tubes
  • Nonsterile cap tubes have unattached, non-looped caps with O-rings
  • Presterilized cap tubes have non-looped caps attached with O-rings
  • Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
  • Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
  • Wide range of colored tubes for easy sample identification
TRUSTED_SUSTAINABILITY
Specifications
Certificeringer/compliance RNase, DNase, pyrogen-free
Lukningsfarve Diverse
Lukningsmateriale Polypropylene
Kapacitet (metrisk) 0.5 mL
Materiale Polypropylene
Format Non-looped
Inkluderer O-ring
Emballage Bag
Sterilitet Ikke-steril
Uddannet No
Lukketype Screw Cap
Bund form Conical
Maks. RCF 20,000 x g