Få mere at vide
Axygen™ 0,5 mL Selvstående rør med skruelåg
Shop alle Axygen Scientific produkter
Sterilitet:
Ikke steril
Ikke-steril
Steril
Lukningsfarve:
Blå
Brun
Diverse
Grøn
Grå
Gul
Hvid
Klar
Orange
Rød
Violet
Pakningsstørrelse:
Pakke med 4000
Pakke med 500
Beskrivelse
For a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping.
- Available as presterilized and nonsterile tubes
- Nonsterile cap tubes have unattached, non-looped caps with O-rings
- Presterilized cap tubes have non-looped caps attached with O-rings
- Threaded O-ring design ensures uniform closure allowing centrifugation up to 20,000 x g
- Maxymum Recovery™ screw cap tubes available as special order items for assorted tubes
- Wide range of colored tubes for easy sample identification
Tekniske data
Tekniske data
| Certificeringer/compliance | RNase, DNase, pyrogen-free |
| Lukningsfarve | Grå |
| Lukningsmateriale | Polypropylene |
| Kapacitet (metrisk) | 0.5 mL |
| Materiale | Polypropylene |
| Format | Non-looped |
| Inkluderer | O-ring |
| Sterilitet | Ikke-steril |
| Uddannet | No |
| Lukketype | Screw Cap |
| Vis mere |