missing translation for 'onlineSavingsMsg'
Få mere at vide

Axitinib Tetrahydro-2H-pyran, TRC

Artikelnummer. 30345291
Change view
Click to view available options
Mængde:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30345291 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30345291 Supplier Toronto Research Chemicals Supplier No. TRCA79513510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Axitinib Tetrahydro-2H-pyran
CAS 885126-35-2
Molekylær formel C27 H26 N4 O2 S
Mængde 10 mg
Synonym Benzamide, N-methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1-(tetrahydro-2H-pyran-2-yl)-1H-indazol-6-yl]thio]- (9CI, ACI), N-Methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1-(tetrahydro-2H-pyran-2-yl)-1H-indazol-6-yl]thio]benzamide (ACI)
InChI formel InChI=1S/C27H26N4O2S/c1-28-27(32)22-9-2-3-10-25(22)34-20-13-14-21-23(15-12-19-8-4-6-16-29-19)30-31(24(21)18-20)26-11-5-7-17-33-26/h2-4,6,8-10,12-16,18,26H,5,7,11,17H2,1H3,(H,28,32)/b15-12+
SMIL CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)nn(C5CCCCO5)c3c2
IUPAC navn N-methyl-2-[1-(oxan-2-yl)-3-[(E)-2-pyridin-2-ylethenyl]indazol-6-yl]sulfanylbenzamide
Molekylvægt (g/mol) 470.586
Formel vægt 470.178
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.