Få mere at vide
AWZ1066S, MedChemExpress
Artikelnummer. 16481087
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16481087 50 mg -
16474828 10 mg -
16467577 5 mg -
16425626 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16481087 Leverandør Medchem Express Leverandørnr. HY114415
MedChemExpress AWZ1066S is a highly specific anti-Wolbachia drug candidate for a short-course treatment of filariasis, with an EC50 of 2.5 nM in cell assay.
AWZ1066S is a highly specific anti-Wolbachia drug candidate for a short-course treatment of filariasis, with an EC50 of 2.5 nM in cell assay.
Tekniske data
Kemisk navn eller materiale AWZ1066S
Fysisk form Solid
CAS 2239272-16-1
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C19H19F3N6O
Til brug med (applikation) COVID-19-immunoregulation
Opløselighedsinformation DMSO : 125 mg/mL (309.11 mM; Need ultrasonic)
SMIL C[C@@H](COCC1)N1C2=NC3=C(C=CC=N3)C(NCC4=CC=CN=C4C(F)(F)F)=N2
Molekylvægt (g/mol) 404.39
Formel vægt 404.39
Procent renhed 98.64%