Få mere at vide
MedChemExpress Aviptadil
Artikelnummer. 16442609
Encompass_Preferred
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
16442609 1 mg
1 options
Artikelnummer. 16442609 Leverandør MedChemExpress Leverandørnr. HYP0012
MedChemExpress Aviptadil is an analog vasoactive intestinal polypeptide (VIP) with potent vasodilatory effects. Aviptadil induces pulmonary vasodilation and inhibits vascular SMCs proliferation, platelet aggregation. Aviptadil can be used for the research of pulmonary fibrosis, pulmonary arterial hypertension (PAH) and SARS-CoV-2 caused respiratory failure, et al.
Tekniske data
Renhed 99.81%
CAS 40077-57-4
Indhold og opbevaring Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Til brug med (applikation) Cancer-programmed cell death
Molekylvægt (g/mol) 3325.8 g/mol
Produkttype Peptides
Mængde 1 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 33.33 mg/mL (10.02 mM∣Need ultrasonic)
Synonym Vasoactive Intestinal Peptide (human, rat, mouse, rabbit, canine, porcine)