Få mere at vide
Avibactam Sodium Salt, TRC
Artikelnummer. 30007491
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007491 5 mg
30009035 10 mg
30003469 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30007491 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7948505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1192491-61-4
Molekylær formel C7 H10 N3 O6 S . Na
Molekylvægt (g/mol) 287.23
Synonym Sulfuric acid, mono[(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester, sodium salt (1:1), Sulfuric acid mono[(1R,2S,5R)-2-(aminocarbonyl)-7-oxo-1,6-diazabicyclo[3.2.1]oct-6-yl] ester monosodium salt, (1R,2S,5R)-7-Oxo-6-sulfooxy-1,6-diazabicyclo[3.2.1]octane-2-carboxamide sodium salt, Avibactam sodium, Avibactam sodium salt
IUPAC navn sodium;[(2S,5R)-2-carbamoyl-7-oxo-1,6-diazabicyclo[3.2.1]octan-6-yl] sulfate
SMIL [Na+].NC(=O)[C@@H]1CC[C@@H]2CN1C(=O)N2OS(=O)(=O)[O-]
Tekniske data
Kemisk navn eller materiale Avibactam sodium salt
Analyt- eller komponentnavne Avibactam sodium salt
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C7H11N3O6S.Na/c8-6(11)5-2-1-4-3-9(5)7(12)10(4)16-17(13,14)15;/h4-5H,1-3H2,(H2,8,11)(H,13,14,15);/q;+1/p-1/t4-,5+;/m1./s1
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 5 mg
Formel vægt 287.0188
Procent renhed >95%