Få mere at vide
Avibactam sodium, MedChemExpress
Artikelnummer. 16462357
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
200 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16462357 50 mg
16486228 10 mg
16476576 100 mg
16418827 5 mg
16401146 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16462357 Leverandør Medchem Express Leverandørnr. HY14879A
MedChemExpress Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
Avibactam sodium (NXL-104) is a covalent and reversible non-β-lactam β-lactamase inhibitor which inhibits β-lactamase TEM-1 and CTX-M-15 with IC50s of 8 nM and 5 nM, respectively.
Tekniske data
Kemisk navn eller materiale Avibactam sodium
CAS 1192491-61-4
Farve White
Renhedsgrad noter Research
Molekylær formel C7H10N3NaO6S
Mængde 50 mg
Synonym NXL-104
Opløselighedsinformation H2O : 50 mg/mL (174.08 mM; Need ultrasonic) ∣DMSO : ≥ 30 mg/mL (104.45 mM)
SMIL O=S(ON1[C@]2([H])CC[C@@H](C(N)=O)[N@@](C2)C1=O)(O[Na])=O
Molekylvægt (g/mol) 287.23
Formel vægt 287.23
Procent renhed 95.0%
Grad Research
Fysisk form Powder
Til brug med (applikation) COVID-19-immunoregulation