missing translation for 'onlineSavingsMsg'
Få mere at vide

Avelestat, TRC

Artikelnummer. 30345027
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30345027 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30345027 Leverandør Toronto Research Chemicals Leverandørnr. TRCA7946605MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Avelestat
CAS 848141-11-7
Molekylær formel C25 H22 F3 N5 O4 S
Mængde 5 mg
InChI formel InChI=1S/C25H22F3N5O4S/c1-15-20(22-9-10-31-32(22)2)12-21(23(34)30-13-17-7-8-19(14-29-17)38(3,36)37)24(35)33(15)18-6-4-5-16(11-18)25(26,27)28/h4-12,14H,13H2,1-3H3,(H,30,34)
SMIL CC1=C(C=C(C(=O)NCc2ccc(cn2)S(=O)(=O)C)C(=O)N1c3cccc(c3)C(F)(F)F)c4ccnn4C
IUPAC navn 6-methyl-5-(2-methylpyrazol-3-yl)-N-[(5-methylsulfonylpyridin-2-yl)methyl]-2-oxo-1-[3-(trifluoromethyl)phenyl]pyridine-3-carboxamide
Molekylvægt (g/mol) 545.53
Formel vægt 545.1345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.