missing translation for 'onlineSavingsMsg'
Learn More

Chemsplash AV MAX Type 4B/5B/6B Coverall
GREENER_CHOICE

AV Max Disposable Coverall is made from a 67 gsm microporous laminated Cat III Type 4B, 5B and 6B fabric.

1640.00 DKK

Specifications

Product Type Cleanroom Coveralls
Gender Unisex
Material Microporous Composite Fabric
Color White
Disposable Single-use
View More Specs

Products 6
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30058147
GREENER_CHOICE
View Documents
Chemsplash
2648.400.004
Small
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058333
GREENER_CHOICE
View Documents
Chemsplash
2648.400.005
Medium
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058158
GREENER_CHOICE
View Documents
Chemsplash
2648.400.006
Large
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058189
GREENER_CHOICE
View Documents
Chemsplash
2648.400.007
X-Large
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058410
GREENER_CHOICE
View Documents
Chemsplash
2648.400.008
2X-Large
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
30058150
GREENER_CHOICE
View Documents
Chemsplash
2648.400.009
3X-Large
1640.00 DKK
Pack of 25
Only null left
Estimated Shipment: 01-08-2025
Add to basket
 
Description

Description

Chemsplash AV Max Disposable Coverall

  • 67GSM Microporous Laminate Fabric
  • Fully Taped Seams
  • Three Piece Hood
  • Elasticated Cuffs with Thumb Loops
  • Elasticated Hood, Back and Ankles
  • Two Way Zip with Zip Flap
  • Silicone and Latex Free
  • Low Linting
  • Anti Static
  • Scores Top Mark of 6 for ISO 16603/16604

Description: The Chemsplash AV Max Disposable Coverall is made from a 67GSM microporous laminated Cat III Type 4B, 5B and 6B fabric. It achieves a top class result of 6 from a possible 6, for resistance to blood borne pathogen testing under ISO 16603 and ISO 16604. All seams are taped offering a stronger and more effective barrier to liquids and particulate matter. Chemsplash Pro +4 fabric is both anti-static to EN1149-5:2008 and is highly resistant to infectious agents under EN14126, recording maximum test scores and complying with resistance to viral and bacterial infection. This coverall is an ideal choice for Pharmaceutical, Cleanroom, Semi-Conductor manufacturing, and Infection Contamination Control.

Specifications

Specifications

Cleanroom Coveralls
Microporous Composite Fabric
Single-use
Elastic
Hooded
Ties
Non-sterile
Yes
Class 100-100,000 (ISO 5-8)
Unisex
White
Elastic
Elastic, Thumb Loop
3-panel
Stitched, Taped
Liquid
None
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.