Få mere at vide
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Kompression Fitting
Til brug med:
1/4 tommer rør yderdiameter
1/8 tommer rør yderdiameter
3/8 tommer rør yderdiameter
4 mm rør OD
6 mm rør OD
Form:
Albue
Lige
T-form
Pakningsstørrelse:
Hver
Beskrivelse
- Available in straight, "T", and elbow shapes for unmatched versatility
- Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
- Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
- Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
- Sterilization through autoclaving or ethylene oxide (ETO)
Tekniske data
Tekniske data
| Autoklaverbar | Autoclavable |
| Materiale | PFA |
| Til brug med | 1/4 tommer rør yderdiameter |
| Emballagemængde | 1 Each |
| Steriliseringsmetode | Autoclave or Ethylene Oxide |
| Form | T-form |
| Produkttype | Compression Fitting |