Learn More
Abeba™ Autoclavable Clogs 39630
Antistatic and autoclavable at 121°C. High slip resistance.
Brand:  Abeba™ 39630/44
468.72 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15611278
  • 480.00 DKK / 1 pair
Only null left
Estimated Shipment: 21-08-2025
Description
  • CE EN ISO 20347:2012 OB A, E SRA
  • Autoclavable, sterilisable special material
  • Orange
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
CE EN ISO 20347:2012 OB A, E SRA
44
39630
Orange
Slip-resistant
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Abeba™ Autoclavable Clogs 39630 > Size: 44