Learn More
Slice™ Auto-Retractable Mini Cutter
Product Code. 16413619
Change view
Click to view available options
Color:
Black/Green
Unit Size:
Each
Product Code. Color Unit Size
16413619 Black/Green Each
1 options
Product Code. 16413619 Supplier Slice™ Supplier No. 10514
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
Forventet forsendelse: 30-11-2026
Features finger-friendly™ safety blade
  • Installed blade type: rounded tip
  • Automatic spring-loaded retraction for added safety
  • Ambidextrous design: Switch blade slots for lefties
  • Finger-friendly™ blade cuts effectively
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • 100 percent zirconium oxide blade
  • Blade edge last up to 11.2x longer than metal
  • Compatible blades: 10404, 10408
  • No-tool blade change
Specifications
For Use With (Equipment) Papers, Bags, Boxes, Tubes
Length (English) 2.44 in.
Length (Metric) 62 mm
Material ABS, POM, Stainless Steel, NdFeB magnet, Zirconium Oxide
Color Black/Green
For Use With (Application) Safety Cutting
Includes 1 rounded tip ceramic blade
No. per Pack 1 Ea.
Weight (English) 0.65 oz.
Weight (Metric) 18.49 g
Width (English) 1.50 in.
Width (Metric) 38 mm
Handle Type Auto-retractable
Product Type Box Cutter, Utility Knife