Learn More
3M™ Aura™ 9300 Series Particulate Respirators
Providing a unique three-panel design for greater comfort and easy communication. 3M™ Aura™ 9300 Series Particulate Respirators cover a range of applications requiring FFP1, FFP2 and FFP3 protection.
Brand:  3M™ 7000034715
Additional Details : Weight : 0.22090kg
 View more versions of this product
Product Code. 11346974
  • 740.00 DKK / Pack of 10
In Stock 7
Add to basket
Description
Description
  • Individually packaged to avoid contamination
  • Soft cover web on inner panel ensures greater comfort
  • Lightweight and foldable
  • Evenly tensioned headbands to relieve strap pressure
  • Centrally positioned cool flow valve reduces heat build-up and makes breathing easier and cooler
Specifications
Specifications
CE Approved to EN 149:2001A1:2009
Flat fold
Polyisoprene strap, Steel staples, Polyurethane nose foam, Aluminum nose clip, Polypropylene filter, Polypropylene valve, Polyisoprene valve diaphragm
White
FFP3
Yes
Dusts and mists found in a wide variety of industrial applications and other work situations
Yes
10 Pack
Particulate Respirator
Three-panel
Yes
Agriculture, cleaning maintenance, demolition
Sculpted
Cup
9332