missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Tocris Bioscience™ AUNP 12

Beskrivelse
AUNP 12 is an immune checkpoint modulator targeting the programmed cell death 1 (PD-1) signaling pathway. It inhibits the interaction between PD-1 and its ligands, PD-L1 and PD-L2. In the mouse splenocyte assay system, AUNP 12 rescues proliferation (EC50 values are 17 and 16 nM for PD-L1 and PD-L2, respectively) and enhances T-cell activity by restoring IFNγ release inhibited by PD-L1 and PD-L2 (EC50 values are 49 nM and 51 nM respectively). In a mouse Renca renal cell carcinoma model, AUNP 12 shows an additive antitumor effect when combined with Cyclophosphamide (Cat. No. 4091).
Tekniske data
Tekniske data
| Gen-id (Entrez) | 5133 |
| Renhed | 95% |
| CAS | 1353563-85-5 |
| Indhold og opbevaring | Store at -20°C |
| Gene Alias | B7-DC, B7H1, B7-H1, Butyrophilin-like Protein, CD273, CD274, CD279, CD279 antigen, hPD-1, IFG, IFI, IFNG, IFNgamma, PD1, PD1hPD-l, PDCD1, PDCD1L1, PDCD1LG1, PDCD1LG2, PDL1, PDL2, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Molekylvægt (g/mol) | M.W. (Observed): 3261.56 |
| Mængde | 1 mg |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?