Learn More
Invitrogen™ Attune™ Xenith™ SIP Shield
Description
The Attune Xenith SIP Shield is for use with the Attune Xenith Flow Cytometer and is a key part of instrument safety, providing a barrier between the user and the sample during acquisition of data. This replacement part is available should any damage occur to the existing part provided with the instrument.
Specifications
Specifications
| Quantity | 1 Ea. |
| For Use With (Equipment) | Attune Xenith Flow Cytometer |
| Product Type | SIP Shield |