Learn More
Attune™ NxT Filter, 569LP
This is a replacement 569 long pass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand:  Invitrogen 100022754
Additional Details : Weight : 0.10000kg
Product Code. 15370637
  • 1384.00 DKK / Each
Estimated Shipment: 08-05-2024
to see stock.
Description
Description
  • Replacement long pass filter
  • Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Specifications
Attune Acoustic Focusing Cytometer
Store at room temperature.
Filter
Attune™