Learn More
Attune™ NxT Filter, 569LP
This is a replacement 569 long pass filter used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer.
Brand: Invitrogen 100022754
Additional Details : Weight : 0.10000kg
Description
- Replacement long pass filter
- Used in the optics path of the Attune™ NxT Acoustic Focusing Cytometer
Specifications
Attune Acoustic Focusing Cytometer | |
Store at room temperature. | |
Filter | |
Attune™ |
For Research Use Only. Not for use in diagnostic procedures.