missing translation for 'onlineSavingsMsg'
Få mere at vide

Atropine N-Oxide Hydrochloride, Mikromol™

Produktkod. 18003460
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Mængde:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Mængde
18003460 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18003460 missing translation for 'mfr' Mikromol missing translation for 'supplierNo' MM0289.190025

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Atropine N-Oxide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifikationer

Kemisk navn eller materiale Atropine N-Oxide Hydrochloride
CAS 4574-60-1
Analyt- eller komponentnavne Atropine N-Oxide Hydrochloride
Forsendelsestilstand Room Temperature
Molekylær formel C17 H23 N O4 . Cl H
Certificeringer/compliance ISO 9001
Synonym Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (1:1), 1αH,5αH-Tropan-3α-ol, (±)-tropate (ester), 8-oxide, hydrochloride (8CI), Benzeneacetic acid, α-(hydroxymethyl)-, (3-endo)-8-methyl-8-oxido-8-azabicyclo[3.2.1]oct-3-yl ester, hydrochloride (9CI), Benzeneacetic acid, α-(hydroxymethyl)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester endo-(±)-, N-oxide, hydrochloride, Atropine N-oxide hydrochloride, Atropine aminoxide hydrochloride, Atropine oxide hydrochloride, Genatropine hydrochloride, Tropinox, Xtro
InChI formel InChI=1S/C17H23NO4.ClH/c1-18(21)13-7-8-14(18)10-15(9-13)22-17(20)16(11-19)12-5-3-2-4-6-12;/h2-6,13-16,19H,7-11H2,1H3;1H/t13-,14+,15+,16-,18?;/m0./s1
SMIL Cl.C[N+]1([O-])[C@@H]2CC[C@H]1C[C@H](C2)OC(=O)[C@@H](CO)c3ccccc3
IUPAC navn [(1R,5S)-8-methyl-8-oxido-8-azoniabicyclo[3.2.1]octan-3-yl] (2R)-3-hydroxy-2-phenylpropanoate;hydrochloride
Molekylvægt (g/mol) 341.83
Mængde 25 mg
Formel vægt 341.1394 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.